Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ern1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023913.2 | NP_076402.1 | 1 - 2 | 180 | 95 | |
GenBank mRNA | AB031332.1 | - | 1 - 2 | 180 | 95 | |
AK018505.1 | - | 1 - 2 | 221 | 95 | ||
AK077041.1 | - | 199 | 95 | |||
AK080709.1 | - | 1 - 2 | 239 | 95 | ||
AK149515.1 | - | 1 - 2 | 195 | 95 | ||
AK170988.1 | - | 1 - 2 | 209 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Mus musculus Inventoried Probe spans exons |