Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2173233 |
BC100447.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2173233 |
AK006193.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2173233 |
AK016415.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2173233 |
NM_026449.3 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
689099 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
230327 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
551765 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391311 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
601339 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
268037 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
415589 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
599249 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
737096 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
774278 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
553353 |
AB009993.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
553353 |
AK148055.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
553353 |
AK157100.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
553353 |
NM_015734.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
341677 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837853 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
266872 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
797893 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
842747 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
529596 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
251537 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428062 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
531616 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
86935 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
180670 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
565438 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
177681 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
653741 |
AK005410.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
84645 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
6395 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
82653 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
137216 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
111853 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P160577 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P154816 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P149033 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2173233 |
BC100447.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2173233 |
AK006193.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2173233 |
AK016415.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2173233 |
NM_026449.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P378006 |
BC100447.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P378006 |
AK006193.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P378006 |
AK016415.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P378006 |
NM_026449.3 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546065 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546044 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546226 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546043 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546052 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546056 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546070 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546064 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546224 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546228 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546062 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546054 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546066 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546229 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546057 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546068 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546059 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546232 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23546220 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05508831 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P160577 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P154816 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01057166 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P149033 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01057146 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P378006 |
BC100447.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P378006 |
AK006193.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P378006 |
AK016415.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P378006 |
NM_026449.3 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P126546 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P154816 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P160577 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P149033 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04438061 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04438065 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04438070 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04738291 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04438072 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546189 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546282 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546261 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546234 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546062 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546132 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546206 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546215 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546043 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546236 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546064 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546244 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546259 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546072 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546279 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546026 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546210 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546233 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546272 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546232 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546033 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546086 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546264 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546035 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546188 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546269 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546211 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546275 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546229 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546220 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546074 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546091 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546155 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546163 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546171 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546256 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546094 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546198 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546142 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546267 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546248 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546179 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546174 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546027 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546250 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546052 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546176 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546260 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546226 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546084 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546245 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546105 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546144 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546207 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546092 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546201 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546124 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546278 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546177 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546147 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546268 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23546097 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546050 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04431463 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546099 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546216 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04334556 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546229 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546044 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04438065 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04896256 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04379212 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546084 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04614885 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546211 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04374111 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546073 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546034 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04604881 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546213 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546228 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04371228 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04158758 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546067 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546052 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04551087 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546101 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546095 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546087 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546036 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546218 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546081 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546070 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04438069 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04374107 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04614887 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546097 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04374108 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546086 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546062 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04334555 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04701702 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04438062 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546224 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546057 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04438073 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546225 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546069 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546066 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546055 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04353687 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04614889 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546082 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546094 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546054 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546071 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546220 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546210 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04374105 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546214 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04374106 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23546226 |
- |
- |