Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125088 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125092 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125096 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125093 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125090 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125094 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125095 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125091 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P420236 |
BC019117.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P420236 |
BC019117.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
200952 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
502200 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
415012 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
511062 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
646426 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
336745 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
918619 |
AK032192.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
918619 |
AK045447.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
918619 |
AK078905.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
918619 |
AK128942.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
918619 |
AK140249.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
918619 |
BC033442.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
918619 |
BC092296.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
918619 |
NM_010604.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
461617 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867549 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
136744 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
40746 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P111152 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P132272 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P420236 |
BC019117.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P420236 |
BC019117.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P420236 |
BC019117.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P420236 |
BC019117.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125015 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125097 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125103 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125012 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125084 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125090 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125009 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125085 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125013 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125082 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125011 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125100 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125094 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125089 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125098 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125088 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125010 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125096 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125007 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125080 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125091 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125081 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125093 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125104 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125006 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24125016 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P111152 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P132272 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P420236 |
BC019117.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P420236 |
BC019117.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P119597 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P111152 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P132272 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04707006 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04692822 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04707003 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04692818 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04707007 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04692820 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04692824 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04707011 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04692821 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125094 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125090 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125091 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125092 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125095 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125096 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125093 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24125088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124995 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124979 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125072 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125043 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124986 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125137 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125079 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125003 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124984 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124947 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125026 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124936 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125053 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125128 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124930 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125038 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124933 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124928 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125035 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124942 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124972 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125040 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124932 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124974 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124981 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124992 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125013 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125032 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124964 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125071 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124975 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125046 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125022 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125096 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125098 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124988 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124977 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125029 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124949 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124938 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24125094 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24124952 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04981385 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125056 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125100 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04707008 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04981384 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125006 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04981374 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04981375 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04981379 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04981398 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04688675 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04981383 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04981394 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04688679 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125104 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125053 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04692822 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04721641 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125037 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04688674 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125040 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04391406 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04502251 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125058 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125093 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125085 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125035 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04688680 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125088 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125024 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125083 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125094 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125101 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04981377 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125046 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125047 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04692818 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04692820 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04504603 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04981372 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04680733 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125071 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04227475 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125078 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125057 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125103 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04981401 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125028 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04391403 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04391400 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04981368 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04680737 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125019 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04369550 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125084 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04981366 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04707010 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125076 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125017 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04680740 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04391390 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04680738 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24125055 |
- |
- |