Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080438.2 | NP_536686.2 | 1 - 2 | 144 | 88 | |
GenBank mRNA | AF362764.1 | - | 1 - 2 | 65 | 88 | |
AK038618.1 | - | 1 - 2 | 510 | 88 | ||
AY230204.1 | - | 1 - 2 | 83 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mus musculus Probe spans exons |