Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008741.4 | NP_032767.1 | 1 - 2 | 173 | 74 | |
GenBank mRNA | AK041338.1 | - | 1 - 2 | 91 | 74 | |
AK049679.1 | - | 1 - 2 | 130 | 74 | ||
AK075612.1 | - | 1 - 2 | 130 | 74 | ||
AK088499.1 | - | 1 - 2 | 165 | 74 | ||
BC018224.1 | - | 1 - 2 | 105 | 4527900 | 74 | |
DN176718.1 | - | 1 - 2 | 173 | 74 | ||
U17259.1 | - | 1 - 2 | 133 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |