Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008894.2 | NP_032920.2 | 8 - 9 | 1146 | 70 | |
GenBank mRNA | AK013901.1 | - | 8 - 9 | 1112 | 70 | |
BC003342.1 | - | 8 - 9 | 1112 | 3257610 | 70 | |
BM215242.2 | - | 3 - 4 | 70 | |||
CB205181.1 | - | 5 - 6 | 682 | 70 | ||
Z72486.1 | - | 8 - 9 | 1107 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |