Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mep1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008586.2 | NP_032612.2 | 6 - 7 | 399 | 85 | |
GenBank mRNA | AK098127.1 | - | 6 - 7 | 399 | 85 | |
BC125627.1 | - | 6 - 7 | 397 | 40130142 | 85 | |
BC145979.1 | - | 6 - 7 | 398 | 40131282 | 85 | |
L15193.1 | - | 7 - 8 | 399 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |