Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009203.3 | NP_033229.3 | 1 - 2 | 723 | 74 | |
GenBank mRNA | AB005451.1 | - | 1 - 2 | 504 | 74 | |
BC035927.1 | - | 659 | 4242354 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |