Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164639.1 | NP_001158111.1 | 3 - 4 | 537 | 108 | |
NM_009289.3 | NP_033315.2 | 3 - 4 | 537 | 108 | ||
GenBank mRNA | AF039574.1 | - | 3 - 4 | 542 | 108 | |
AF112855.1 | - | 3 - 4 | 426 | 108 | ||
AK029491.1 | - | 3 - 4 | 404 | 108 | ||
AK122218.1 | - | 3 - 4 | 878 | 108 | ||
BC131674.1 | - | 3 - 4 | 438 | 40110915 | 108 | |
BC131675.1 | - | 3 - 4 | 438 | 40110921 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |