Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161623.1 | NP_001155095.1 | 15 - 16 | 1491 | 91 | |
NM_009862.2 | NP_033992.2 | 17 - 18 | 1868 | 91 | ||
GenBank mRNA | AF081536.1 | - | 17 - 18 | 1868 | 91 | |
AF081537.1 | - | 16 - 17 | 1567 | 91 | ||
AF081538.1 | - | 16 - 17 | 1567 | 91 | ||
AF081539.1 | - | 4 - 5 | 452 | 91 | ||
AF098068.1 | - | 16 - 17 | 1614 | 91 | ||
AJ223729.1 | - | 16 - 17 | 1588 | 91 | ||
AK147013.1 | - | 16 - 17 | 1585 | 91 | ||
AK167850.1 | - | 15 - 16 | 1479 | 91 | ||
BC006856.1 | - | 16 - 17 | 1583 | 91 | ||
BC028635.1 | - | 16 - 17 | 1621 | 1262927 | 91 | |
CO040149.1 | - | 3 - 4 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |