Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009867.2 | NP_033997.1 | 8 - 9 | 1318 | 100 | |
GenBank mRNA | D14888.1 | - | 8 - 9 | 1228 | 100 | |
X69966.1 | - | 8 - 9 | 1174 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Inventoried Probe spans exons |