Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011355.1 | NP_035485.1 | 3 - 4 | 485 | 91 | |
GenBank mRNA | AK142246.1 | - | 3 - 4 | 502 | 91 | |
AK153524.1 | - | 3 - 4 | 516 | 91 | ||
AK154945.1 | - | 3 - 4 | 537 | 91 | ||
AK155085.1 | - | 3 - 4 | 521 | 91 | ||
BC003815.1 | - | 3 - 4 | 403 | 3600260 | 91 | |
L03215.1 | - | 3 - 4 | 382 | 91 | ||
M32370.1 | - | 3 - 4 | 465 | 91 | ||
M38252.1 | - | 3 - 4 | 485 | 91 | ||
X17463.1 | - | 3 - 4 | 1173 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |