Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199003.1 | NP_001185932.1 | 13 - 14 | 1286 | 94 | |
NM_011880.3 | NP_036010.2 | 13 - 14 | 1286 | 94 | ||
GenBank mRNA | AF011360.1 | - | 13 - 14 | 1131 | 94 | |
BC051133.1 | - | 13 - 14 | 1241 | 6588901 | 94 | |
DQ139288.1 | - | 1 - 2 | 64 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |