Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016759.4 | NP_058039.1 | 8 - 9 | 1225 | 99 | |
GenBank mRNA | AK002856.1 | - | 4 - 5 | 523 | 99 | |
BC022745.1 | - | 5 - 6 | 1057 | 99 | ||
U73941.1 | - | 8 - 9 | 952 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |