Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252614.1 | NP_001239543.1 | 10 - 11 | 1683 | 88 | |
NM_001252615.1 | NP_001239544.1 | 4 - 5 | 757 | 88 | ||
NM_001252616.1 | NP_001239545.1 | 3 - 4 | 777 | 88 | ||
NM_016893.5 | NP_058589.2 | 9 - 10 | 1594 | 88 | ||
NR_045554.1 | 9 - 10 | 2013 | 88 | |||
GenBank mRNA | AB025198.1 | - | 6 - 7 | 1283 | 88 | |
AK048520.1 | - | 9 - 10 | 1732 | 88 | ||
AK051811.1 | - | 9 - 10 | 1551 | 88 | ||
AK134537.1 | - | 9 - 10 | 1687 | 88 | ||
BC010666.1 | - | 10 - 11 | 1662 | 3594582 | 88 | |
EF591874.1 | - | 4 - 5 | 757 | 88 | ||
EF591875.1 | - | 3 - 4 | 777 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |