Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008446.2 | NP_032472.2 | 20 - 21 | 2797 | 105 | |
GenBank mRNA | AK135430.1 | - | 20 - 21 | 2291 | 105 | |
AK171938.1 | - | 20 - 21 | 2709 | 105 | ||
BC050946.1 | - | 20 - 21 | 2735 | 6822283 | 105 | |
BC054537.1 | - | 20 - 21 | 2719 | 105 | ||
D12646.1 | - | 20 - 21 | 3124 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |