Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191023.1 | NP_001177952.1 | 7 - 8 | 1038 | 101 | |
NM_001191025.1 | NP_001177954.1 | 7 - 8 | 987 | 101 | ||
NM_001191026.1 | NP_001177955.1 | 8 - 9 | 1014 | 101 | ||
NM_001191027.1 | NP_001177956.1 | 7 - 8 | 954 | 101 | ||
NM_010063.4 | NP_034193.2 | 8 - 9 | 1047 | 101 | ||
GenBank mRNA | AF063229.1 | - | 8 - 9 | 881 | 101 | |
AF063230.1 | - | 7 - 8 | 821 | 101 | ||
AK158589.1 | - | 8 - 9 | 1015 | 101 | ||
BC051992.1 | - | 8 - 9 | 775 | 5696629 | 101 | |
GU992206.1 | - | 7 - 8 | 779 | 101 | ||
GU992207.1 | - | 7 - 8 | 695 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |