Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011920.3 | NP_036050.1 | 3 - 4 | 718 | 93 | |
GenBank mRNA | AA008579.1 | - | 344 | 93 | ||
AF140218.1 | - | 2 - 3 | 281 | 93 | ||
AK142528.1 | - | 3 - 4 | 364 | 93 | ||
BC053730.1 | - | 3 - 4 | 730 | 30064535 | 93 | |
CO039662.1 | - | 1 - 2 | 93 | |||
KF662864.1 | - | 3 - 4 | 315 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |