Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uchl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159866.1 | NP_001153338.1 | 5 - 6 | 560 | 74 | |
NM_019562.2 | NP_062508.2 | 5 - 6 | 560 | 74 | ||
GenBank mRNA | AF148447.1 | - | 5 - 6 | 503 | 74 | |
AF175903.1 | - | 5 - 6 | 512 | 74 | ||
AK007860.1 | - | 4 - 5 | 374 | 74 | ||
AK011117.1 | - | 5 - 6 | 527 | 74 | ||
AK017925.1 | - | 5 - 6 | 558 | 74 | ||
BC006891.1 | - | 5 - 6 | 513 | 2650799 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |