Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252443.1 | NP_001239372.1 | 4 - 5 | 746 | 75 | |
NM_019678.3 | NP_062652.1 | 5 - 6 | 893 | 75 | ||
GenBank mRNA | AK008549.1 | - | 5 - 6 | 669 | 75 | |
AK088864.1 | - | 4 - 5 | 564 | 75 | ||
AK158539.1 | - | 5 - 6 | 759 | 75 | ||
AK167475.1 | - | 5 - 6 | 757 | 75 | ||
BC024638.1 | - | 5 - 6 | 671 | 4015300 | 75 | |
U94662.1 | - | 5 - 6 | 795 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |