Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031198.3 | NP_112475.1 | 1 - 2 | 180 | 69 | |
GenBank mRNA | AF077742.1 | - | 1 - 2 | 180 | 69 | |
AK137141.1 | - | 1 - 2 | 145 | 69 | ||
AK156327.1 | - | 1 - 2 | 93 | 69 | ||
AK157426.1 | - | 1 - 2 | 180 | 69 | ||
AK159579.1 | - | 1 - 2 | 113 | 69 | ||
BC052832.1 | - | 1 - 2 | 143 | 69 | ||
BC064770.1 | - | 1 - 2 | 131 | 69 | ||
BC098494.1 | - | 1 - 2 | 65 | 6431607 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |