Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ocln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008756.2 | NP_032782.1 | 6 - 7 | 1427 | 139 | |
GenBank mRNA | AK019880.1 | - | 6 - 7 | 1379 | 139 | |
AK033532.1 | - | 6 - 7 | 1360 | 139 | ||
AK042576.1 | - | 6 - 7 | 1427 | 139 | ||
AK145798.1 | - | 4 - 5 | 586 | 139 | ||
BC138679.1 | - | 6 - 7 | 1410 | 8861700 | 139 | |
BC138680.1 | - | 6 - 7 | 1410 | 8861701 | 139 | |
BC145371.1 | - | 6 - 7 | 1425 | 9053929 | 139 | |
U49185.1 | - | 6 - 7 | 1470 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |