Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030251.3 | NP_084527.2 | 7 - 8 | 756 | 72 | |
GenBank mRNA | AB053477.1 | - | 7 - 8 | 687 | 72 | |
AK049100.1 | - | 7 - 8 | 672 | 72 | ||
AK088437.1 | - | 7 - 8 | 721 | 72 | ||
AK089474.1 | - | 5 - 6 | 1120 | 72 | ||
AK155432.1 | - | 2 - 3 | 2178 | 72 | ||
AK155505.1 | - | 2 - 3 | 2178 | 72 | ||
AK155644.1 | - | 2 - 3 | 2178 | 72 | ||
BC003234.1 | - | 7 - 8 | 700 | 3483893 | 72 | |
CO804154.1 | - | 2 - 3 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |