Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130884.4 | NP_570954.1 | 8 - 9 | 798 | 104 | |
GenBank mRNA | AK215024.1 | - | 2 - 3 | 187 | 104 | |
AK216020.1 | - | 3 - 4 | 336 | 104 | ||
AY035212.1 | - | 8 - 9 | 779 | 104 | ||
BC009022.1 | - | 8 - 9 | 787 | 3257667 | 104 | |
CN456483.1 | - | 3 - 4 | 346 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |