Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139151.4 | NP_631890.3 | 7 - 8 | 871 | 71 | |
GenBank mRNA | AB041554.1 | - | 5 - 6 | 71 | ||
AK028080.1 | - | 7 - 8 | 840 | 71 | ||
AK043812.1 | - | 7 - 8 | 839 | 71 | ||
AK220494.1 | - | 2144 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |