Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139227.4 | NP_631973.3 | 8 - 9 | 1575 | 85 | |
GenBank mRNA | AF455111.1 | - | 7 - 8 | 1315 | 85 | |
AK046679.1 | - | 8 - 9 | 1576 | 85 | ||
BC035459.1 | - | 7 - 8 | 1228 | 85 | ||
BC137846.1 | - | 8 - 9 | 1496 | 8860865 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |