Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113554.1 | NP_001107026.1 | 8 - 9 | 1827 | 91 | |
NM_026149.4 | NP_080425.3 | 8 - 9 | 1830 | 91 | ||
GenBank mRNA | AF521132.1 | - | 8 - 9 | 1828 | 91 | |
AK014996.2 | - | 8 - 9 | 1830 | 91 | ||
AK077673.1 | - | 8 - 9 | 1412 | 91 | ||
BC031583.1 | - | 8 - 9 | 1396 | 4952090 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |