Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028064.2 | NP_082340.1 | 6 - 7 | 1267 | 65 | |
GenBank mRNA | AK005535.1 | - | 6 - 7 | 1307 | 65 | |
AK146977.1 | - | 6 - 7 | 1308 | 65 | ||
AK147107.1 | - | 6 - 7 | 1286 | 65 | ||
AK154089.1 | - | 6 - 7 | 1786 | 65 | ||
AK155467.1 | - | 6 - 7 | 1786 | 65 | ||
BC023498.1 | - | 6 - 7 | 1267 | 5011088 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |