Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdh1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029696.4 | NP_083972.3 | 5 - 6 | 936 | 100 | |
GenBank mRNA | AK007262.1 | - | 1 - 2 | 61 | 100 | |
BC050786.1 | - | 5 - 6 | 936 | 6703279 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |