Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdh1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029696.4 | NP_083972.3 | 9 - 10 | 1433 | 108 | |
GenBank mRNA | AK007262.1 | - | 5 - 6 | 558 | 108 | |
BC050786.1 | - | 9 - 10 | 1433 | 6703279 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |