Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007771.3 | NP_031797.1 | 7 - 8 | 1723 | 89 | |
GenBank mRNA | AB000777.1 | - | 7 - 8 | 1159 | 89 | |
AF156986.1 | - | 7 - 8 | 1458 | 89 | ||
AK133818.1 | - | 7 - 8 | 1735 | 89 | ||
AK162460.1 | - | 7 - 8 | 1723 | 89 | ||
BC022174.1 | - | 7 - 8 | 1494 | 4036758 | 89 | |
BC085499.1 | - | 7 - 8 | 1723 | 6813367 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |