Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009849.2 | NP_033979.2 | 2 - 3 | 334 | 57 | |
GenBank mRNA | AF042811.1 | - | 2 - 3 | 258 | 57 | |
AK002553.1 | - | 2 - 3 | 306 | 57 | ||
AK172281.1 | - | 2 - 3 | 282 | 57 | ||
AY376711.1 | - | 2 - 3 | 267 | 57 | ||
BC011241.1 | - | 2 - 3 | 266 | 3582550 | 57 | |
U91511.1 | - | 2 - 3 | 301 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |