Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dazl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277863.1 | NP_001264792.1 | 8 - 9 | 893 | 142 | |
NM_010021.5 | NP_034151.3 | 9 - 10 | 944 | 142 | ||
GenBank mRNA | AK014854.1 | - | 9 - 10 | 924 | 142 | |
AK033183.1 | - | 9 - 10 | 924 | 142 | ||
AK160855.1 | - | 9 - 10 | 803 | 142 | ||
U38690.1 | - | 9 - 10 | 1059 | 142 | ||
U46694.1 | - | 9 - 10 | 840 | 142 | ||
X95724.1 | - | 9 - 10 | 833 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |