Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012001.2 | NP_036131.1 | 2 - 3 | 321 | 76 | |
GenBank mRNA | AF071314.1 | - | 2 - 3 | 190 | 76 | |
AK003752.1 | - | 2 - 3 | 183 | 76 | ||
AK009955.1 | - | 2 - 3 | 193 | 76 | ||
AK162763.1 | - | 2 - 3 | 187 | 76 | ||
AK167833.1 | - | 2 - 3 | 207 | 76 | ||
BC116800.1 | - | 2 - 3 | 175 | 40126119 | 76 | |
BC116826.1 | - | 2 - 3 | 175 | 40126145 | 76 | |
CX203220.1 | - | 2 - 3 | 321 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |