Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200007152.mm.1 |
NM_013747.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007152.mm.1 |
NM_013747.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007152.mm.1 |
TC1200001017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007149.mm.1 |
NM_001199004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007149.mm.1 |
NM_001199004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007149.mm.1 |
TC1200001017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007153.mm.1 |
NM_001199004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007153.mm.1 |
NM_001199004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007153.mm.1 |
NM_013747.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007153.mm.1 |
NM_013747.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007153.mm.1 |
TC1200001017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007154.mm.1 |
NM_001199004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007154.mm.1 |
NM_001199004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007154.mm.1 |
NM_013747.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007154.mm.1 |
NM_013747.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007154.mm.1 |
TC1200001017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007150.mm.1 |
NM_001199004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007150.mm.1 |
NM_001199004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007150.mm.1 |
TC1200001017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007151.mm.1 |
TC1200001017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007155.mm.1 |
NM_001199004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007155.mm.1 |
NM_001199004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007155.mm.1 |
NM_013747.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007155.mm.1 |
NM_013747.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007155.mm.1 |
TC1200001017.mm.1 |
MTA TC |