Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps8l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133867.2 | NP_598628.1 | 1 - 2 | 138 | 88 | |
GenBank mRNA | AK136584.1 | - | 1 - 2 | 134 | 88 | |
AK167630.1 | - | 1 - 2 | 125 | 88 | ||
AY074932.1 | - | 1 - 2 | 154 | 88 | ||
BC014734.1 | - | 1 - 2 | 126 | 4218079 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |