Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138592.4 | NP_613058.1 | 10 - 11 | 1445 | 136 | |
GenBank mRNA | AK010443.1 | - | 10 - 11 | 1455 | 136 | |
AK018595.1 | - | 10 - 11 | 1446 | 136 | ||
AK033105.1 | - | 10 - 11 | 1442 | 136 | ||
AK045062.1 | - | 10 - 11 | 1442 | 136 | ||
AK088354.1 | - | 10 - 11 | 1457 | 136 | ||
AK165625.1 | - | 10 - 11 | 1426 | 136 | ||
AK167668.1 | - | 10 - 11 | 1433 | 136 | ||
BC026983.1 | - | 10 - 11 | 1420 | 5365862 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |