Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mios | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145374.2 | NP_663349.2 | 2 - 3 | 247 | 86 | |
GenBank mRNA | BC020002.1 | - | 2 - 3 | 245 | 3584335 | 86 |
BU510538.1 | - | 2 - 3 | 97 | 86 | ||
BY064617.1 | - | 2 - 3 | 247 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |