Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145482.3 | NP_663457.2 | 2 - 3 | 325 | 70 | |
GenBank mRNA | AY037804.1 | - | 2 - 3 | 325 | 70 | |
BC029101.1 | - | 2 - 3 | 222 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |