Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053177.1 | NP_444407.1 | 8 - 9 | 1059 | 65 | |
GenBank mRNA | AF302009.1 | - | 8 - 9 | 1056 | 65 | |
AK028385.1 | - | 8 - 9 | 1036 | 65 | ||
AK034471.1 | - | 8 - 9 | 1070 | 65 | ||
AK038594.1 | - | 8 - 9 | 1072 | 65 | ||
AK049606.1 | - | 8 - 9 | 1060 | 65 | ||
BC005651.1 | - | 8 - 9 | 1059 | 3257440 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |