Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gys2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145572.2 | NP_663547.2 | 14 - 15 | 2115 | 98 | |
GenBank mRNA | AK138992.1 | - | 14 - 15 | 1823 | 98 | |
AK149391.1 | - | 14 - 15 | 1840 | 98 | ||
BC021322.1 | - | 14 - 15 | 1976 | 5051685 | 98 | |
BC158081.1 | - | 14 - 15 | 1976 | 9007373 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |