Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146150.2 | NP_666262.2 | 10 - 11 | 1426 | 97 | |
GenBank mRNA | AK031548.1 | - | 10 - 11 | 1426 | 97 | |
AK040377.1 | - | 9 - 10 | 1657 | 97 | ||
AK166470.1 | - | 8 - 9 | 1425 | 97 | ||
BC023786.1 | - | 9 - 10 | 804 | 97 | ||
BC026832.1 | - | 10 - 11 | 1476 | 4486176 | 97 | |
BC029977.1 | - | 1 - 2 | 62 | 97 | ||
BC036128.1 | - | 10 - 11 | 1544 | 4971791 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |