Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24153010 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24152942 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24153013 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24153009 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24152943 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24152941 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24153011 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24152945 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2048119 |
BC025599.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2048119 |
NM_146257.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
122798 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
219351 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
902990 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
403523 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
76719 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325076 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
251711 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
857077 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
372309 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
802878 |
AF067060.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
802878 |
BC125501.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
802878 |
BC125503.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
802878 |
NM_153523.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
494213 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
413472 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
564417 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
179492 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P142297 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P160666 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2048119 |
BC025599.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2048119 |
NM_146257.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P1959 |
BC025599.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P1959 |
NM_146257.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24152944 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24152936 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24152933 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24152930 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24152941 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24152943 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24152946 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24152939 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24152945 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24152934 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P142297 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P160666 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05773347 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P1959 |
BC025599.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P1959 |
NM_146257.2 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P118302 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P160666 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P142297 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04773116 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04773120 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04773124 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04773121 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24152942 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24153010 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24153013 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24152941 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24152943 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24152945 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24153011 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24153009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152983 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152973 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152959 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152933 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152930 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152939 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152961 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152916 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152979 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153005 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152909 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152969 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152907 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152934 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152923 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152925 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153027 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152972 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153001 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152941 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153004 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152955 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152936 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152944 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152976 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152921 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152917 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152943 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153008 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24153014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152910 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24152905 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152952 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24153024 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04350841 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04682128 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152927 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152918 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152958 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24153028 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04773116 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152954 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152930 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04773120 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152957 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04166356 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24153021 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04980748 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152955 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04265053 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152947 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24153027 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152910 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04809713 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24153020 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04790262 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04809710 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152911 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152923 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04773121 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04773124 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152909 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24152920 |
- |
- |