Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aatk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198785.1 | NP_001185714.1 | 6 - 7 | 843 | 64 | |
NM_001198787.1 | NP_001185716.1 | 6 - 7 | 615 | 64 | ||
NM_007377.4 | NP_031403.2 | 6 - 7 | 1126 | 64 | ||
GenBank mRNA | AB093253.2 | - | 6 - 7 | 1126 | 64 | |
AB288871.1 | - | 6 - 7 | 745 | 64 | ||
AF011908.1 | - | 6 - 7 | 582 | 64 | ||
AK036705.1 | - | 6 - 7 | 617 | 64 | ||
AK147239.1 | - | 3 - 4 | 636 | 64 | ||
AK155100.1 | - | 6 - 7 | 617 | 64 | ||
AY236858.1 | - | 6 - 7 | 621 | 64 | ||
AY236859.1 | - | 6 - 7 | 648 | 64 | ||
BC042837.1 | - | 6 - 7 | 1148 | 64 | ||
BC052323.1 | - | 6 - 7 | 1148 | 64 | ||
BC080846.1 | - | 6 - 7 | 697 | 6856191 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |