Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164610.1 | NP_001158082.1 | 13 - 14 | 1329 | 62 | |
NM_029945.3 | NP_084221.2 | 14 - 15 | 1416 | 62 | ||
GenBank mRNA | AK019466.2 | - | 14 - 15 | 1417 | 62 | |
AK031685.1 | - | 13 - 14 | 1328 | 62 | ||
AK035892.1 | - | 13 - 14 | 1321 | 62 | ||
AK036055.1 | - | 13 - 14 | 1321 | 62 | ||
AK036380.1 | - | 13 - 14 | 1325 | 62 | ||
AK129354.1 | - | 14 - 15 | 1393 | 62 | ||
AK138720.1 | - | 13 - 14 | 1303 | 62 | ||
AK164456.1 | - | 11 - 12 | 1794 | 62 | ||
BC026767.1 | - | 13 - 14 | 1302 | 5137008 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |