Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163570.1 | NP_001157042.1 | 6 - 7 | 699 | 75 | |
NM_027148.3 | NP_081424.3 | 6 - 7 | 687 | 75 | ||
GenBank mRNA | AJ505005.1 | - | 6 - 7 | 347 | 75 | |
AK009584.1 | - | 6 - 7 | 359 | 75 | ||
AK169719.1 | - | 4 - 5 | 290 | 75 | ||
AK220227.1 | - | 6 - 7 | 687 | 75 | ||
BC059089.1 | - | 6 - 7 | 716 | 6509820 | 75 | |
BC067250.1 | - | 5 - 6 | 330 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |