Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172439.3 | NP_766027.2 | 1 - 2 | 177 | 86 | |
GenBank mRNA | AK034272.1 | - | 1 - 2 | 178 | 86 | |
AK136908.1 | - | 1 - 2 | 162 | 86 | ||
AK170114.1 | - | 1 - 2 | 143 | 86 | ||
AK171029.1 | - | 1 - 2 | 138 | 86 | ||
BC022915.1 | - | 1 - 2 | 130 | 86 | ||
BC051452.1 | - | 1 - 2 | 147 | 86 | ||
BC131634.1 | - | 1 - 2 | 172 | 40096940 | 86 | |
BC131635.1 | - | 1 - 2 | 169 | 40096943 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |