Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mb21d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173386.5 | NP_775562.2 | 3 - 4 | 1234 | 109 | |
NR_125721.1 | 3 - 4 | 1278 | 109 | |||
GenBank mRNA | AK030292.1 | - | 3 - 4 | 1236 | 109 | |
AK054330.1 | - | 3 - 4 | 1168 | 109 | ||
AK078662.1 | - | 3 - 4 | 1145 | 109 | ||
AK145268.1 | - | 3 - 4 | 1174 | 109 | ||
AK154165.1 | - | 3 - 4 | 1203 | 109 | ||
BC044854.1 | - | 3 - 4 | 938 | 109 | ||
BC052196.1 | - | 3 - 4 | 1144 | 6510706 | 109 | |
BC145651.1 | - | 3 - 4 | 1204 | 40130954 | 109 | |
BC145653.1 | - | 3 - 4 | 1204 | 40130956 | 109 | |
KC294567.1 | - | 3 - 4 | 1075 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |