Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175092.3 | NP_780301.1 | 1 - 2 | 401 | 67 | |
GenBank mRNA | AK038837.1 | - | 169 | 67 | ||
AK172212.1 | - | 1 - 2 | 169 | 67 | ||
BC096597.1 | - | 1 - 2 | 191 | 4983233 | 67 | |
BC132355.1 | - | 233 | 40130632 | 67 | ||
BC132357.1 | - | 233 | 40130634 | 67 | ||
CK617811.1 | - | 1 - 2 | 408 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |