Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175314.3 | NP_780523.2 | 16 - 17 | 2367 | 106 | |
GenBank mRNA | AK030699.1 | - | 3 - 4 | 477 | 106 | |
AK173146.1 | - | 3 - 4 | 487 | 106 | ||
BC068142.1 | - | 2 - 3 | 406 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |